Learn More
enQuireBio™ Recombinant Human ASGR2 Protein
A cDNA sequence encoding the ASGR2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11085-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
ASGR2 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ASGR2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 30% glycerol and 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQSEGHRG AQLQAELRSL KEAFSNFSSS TLTEVQAIST HGGSVGDKIT SLGAKLEKQQ QDLKADHDAL LFHLKHFPVD LRFVACQMEL LHSNGSQRTC CPVNWVEHQG SCYWFSHSGK AWAEAEKYCQ LENAHLVVIN SWEEQKFIVQ HTNPFNTWIG LTDSDGSWKW VDGTDYRHNY KNWAVTQPDN WHGHELGGSE DCVEVQPDGR WNDDFCLQVY RWVCEKRRNA TGEVA |