missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ASPRV1 Protein

A cDNA sequence encoding the ASPRV1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11091-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15996108

  • 205.00€ / 20µg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

ASPRV1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ASPRV1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSMGKGYY LKGKIGKVPV RFLVDSGAQV SVVHPNLWEE VTDGDLDTLQ PFENVVKVAN GAEMKILGVW DTAVSLGKLK LKAQFLVANA SAEEAIIGTD VLQDHNAILD FEHRTCTLKG KKFRLLPVGG SLEDEFDLE
Vidéos
FDS
Documents
Certificats
Promotions

Promotions