Learn More
enQuireBio™ Recombinant Human ATP1B1 Protein
A cDNA sequence encoding the ATP1B1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11104-HIS-EC-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
ATP1B1 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ATP1B1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFKPTYQ DRVAPPGLTQ IPQIQKTEIS FRPNDPKSYE AYVLNIVRFL EKYKDSAQRD DMIFEDCGDV PSEPKERGDF NHERGERKVC RFKLEWLGNC SGLNDETYGY KEGKPCIIIK LNRVLGFKPK PPKNESLETY PVMKYNPNVL PVQCTGKRDE DKDKVGNVEY FGLGNSPGFP LQYYPYYGKL LQPKYLQPLL AVQFTNLTMD TEIRIECKAY GENIGYSEKD RFQGRFDVKI EVKS |