Learn More
enQuireBio™ Recombinant Human ATP1B3 Protein
A cDNA sequence encoding the ATP1B3 was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP11106-2ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15916168
  • 131.00€ / 2µg
Expédition estimée: 10-05-2024
Spécification
ATP1B3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSTMWVMLQ TLNDEVPKYR DQIPSPGLMV FPKPVTALEY TFSRSDPTSY AGYIEDLKKF LKPYTLEEQK NLTVCPDGAL FEQKGPVYVA CQFPISLLQA CSGMNDPDFG YSQGNPCILV KMNRIIGLKP EGVPRIDCVS KNEDIPNVAV YPHNGMIDLK YFPYYGKKLH VGYLQPLVAV QVSFAPNNTG KEVTVECKID GSANLKSQDD RDKFLGRVMF KITARA
Greater than 80.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ATP1B3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.15M NaCl and 1mM DTT.