missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP5C1 Protein
A cDNA sequence encoding the ATP5C1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11107-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
ATP5C1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSATLKDIT RRLKSIKNIQ KITKSMKMVA AAKYARAERE LKPARIYGLG SLALYEKADI KGPEDKKKHL LIGVSSDRGL CGAIHSSIAK QMKSEVATLT AAGKEVMLVG IGDKIRGILY RTHSDQFLVA FKEVGRKPPT FGDASVIALE LLNSGYEFDE GSIIFNKFRS VISYKTEEKP IFSLNTVASA DSMSIYDDID ADVLQNYQEY NLANIIYYSL KESTTSEQSA RMTAMDNASK NASEMIDKLT LTFNRTRQAV ITKELIEIIS GAAALD | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ATP5C1 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M Urea and 10% glycerol. |