Learn More
enQuireBio™ Recombinant Human BMF / Bcl2 modifying factor Protein
A DNA sequence encoding the Homo sapiens (Human) BMF / Bcl2 modifying factor, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7962-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001003940.1 | |
BMF / Bcl2 modifying factor Protein | |
1 mg | |
Not determined. | |
Full Length of BC069505 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
90427 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
BMF | |
Recombinant Protein | |
E. coli | |
MEPSQCVEEL EDDVFQPEDG EPVTQPGSLL SADLFAQSLL DCPLSRLQLF PLTHCCGPGL RPTSQEDKAT QTLSPASPSP GVMLPCGVTE EPQRLFYGNA GYRLPLPASF PAVLPIGEQP PEGQWQHQAE VQIARKLQCI ADQFHRLHVQ QHQQNQNRVW WQILLFLHNL ALNGEENRNG AGPR |