missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human BOD1 GST (N-Term) Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
10 μg
25 μg
Conditionnement:
10µg
25µg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spécification
Spécification
| À utiliser avec (application) | Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification |
| Formule | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Identification génétique (Entrez) | 91272 |
| Poids moléculaire | MolecularWeight-theroretical: 45.6 kDa |
| Quantité | 25 μg |
| Conditions de stockage | Store at -80°C. Avoid freeze/thaw cycles. |
| Alias de gène | biorientation defective 1, Biorientation defective protein 1, biorientation of chromosomes in cell division 1, biorientation of chromosomes in cell division protein 1, FAM44B, family with sequence similarity 44, member B, Protein FAM44B |
| Symbole de gène(s) | BOD1 |
| Pureté ou qualité | >80% by SDS-PAGE and Coomassie blue staining |
| Protéine | BOD1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu