Learn More
enQuireBio™ Recombinant Human C6ORF108 Protein
A cDNA sequence encoding the C6ORF108 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11241-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
C6ORF108 Protein | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The C6ORF108 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8), 1mM DTT, and 10% glycerol. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAAMVPGRS ESWERGEPGR PALYFCGSIR GGREDRTLYE RIVSRLRRFG TVLTEHVAAA ELGARGEEAAGGDRLIHEQD LEWLQQADVV VAEVTQPSLG VGYELGRAVA FNKRILCLFR PQSGRVLSAM IRGAADGSRF QVWDYEEGEV EALLDRYFEA DPPGQVAASP DPTT |