missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C9 Protein

A cDNA sequence encoding the C9 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11243-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15996588

  • 205.00€ / 10µg
Expédition estimée: 09-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

C9 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
C9 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQYTTSYD PELTESSGSA SHIDCRMSPW SEWSQCDPCL RQMFRSRSIE VFGQFNGKRC TDAVGDRRQC VPTEPCEDAE DDCGNDFQCS TGRCIKMRLR CNGDNDCGDF SDEDDCESEP RPPCRDRVVE ESELARTAGY GINILGMDPL STPFDNEFYN GLCNRDRDGN TLTYYRRPWN VASLIYETKG EKNFRTEHYE EQIEAFKSII QEKTSNFNAA ISLKFTPTET NKAEQCCEET ASSISLHGKG SFRFSYSKNE TYQLFLSYSS KKEKMFLHVK GEIHLGRFVM RNRDVVLTTT FVDDIKALPT TYEKGEYFAF LETYGTHYSS SGSLGGLYEL IYVLDKASMK RKGVELKDIK RCLGYHLDVS LAFSEISVGA EFNKDDCVKR GEGRAVNITS ENLIDDVVSL IRGGTRKYAF ELKEKLLRGT VIDVTDFVNW ASSINDAPVL ISQKLSPIYN LVPVKMKNAH LKKQNLERAI EDYINEFSVR KCHTCQNGGT VILMDGKCLC ACPFKFEGIA CEISKQKISE GLPALEFPNE K
Vidéos
FDS
Documents
Certificats
Promotions

Promotions