Learn More
enQuireBio™ Recombinant Human Calpain small subunit 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Calpain small subunit 1, was expressed in the hosts and tags indicated.
Marque:  enQuireBio™ QP5758-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15977539
  • 476.00€ / 100µg
Expédition estimée: 10-07-2023
Spécification
NP_001003962.1
Calpain small subunit 1 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
826
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CAPNS1
Recombinant Protein
E. coli
MFLVNSFLKG GGGGGGGGGG LGGGLGNVLG GLISGAGGGG GGGGGGGGGG GGGGGGTAMR ILGGVISAIS EAAAQYNPEP PPPRTHYSNI EANESEEVRQ FRRLFAQLAG DDMEVSATEL MNILNKVVTR HPDLKTDGFG IDTCRSMVAV MDSDTTGKLG FEEFKYLWNN IKRWQAIYKQ FDTDRSGTIC SSELPGAFEA AGFHLNEHLY NMIIRRYSDE SGNMDFDNFI SCLVRLDAMF RAFKSLDKDG TGQIQVNIQE WLQLTMYS