Learn More
enQuireBio™ Recombinant Human Cancer / testis antigen 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Cancer/testis antigen 2, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5883-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_066274.2 | |
Cancer / testis antigen 2 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
30848 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CTAG2 | |
Recombinant Protein | |
E. coli | |
MQAEGQGTGG STGDADGPGG PGIPDGPGGN AGGPGEAGAT GGRGPRGAGA ARASGPRGGA PRGPHGGAAS AQDGRCPCGA RRPDSRLLQL HITMPFSSPM EAELVRRILS RDAAPLPRPG AVLKDFTVSG NLLFMSVRDQ DREGAGRMRV VGWGLGSASP EGQKARDLRT PKHKVSEQRP GTPGPPPPEG AQGDGCRGVA FNVMFSAPHI |