missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL3 / Mip1a Protein
Recombinant Protein
Marque: enQuireBio™ QP5274-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_002974.1 | |
CCL3 / Mip1a Protein | |
Research Use Only | |
CCL3 | |
Recombinant Protein | |
E. coli | |
ASLAADTPTA CCFSYTSRQI PQNFIADYFE TSSQCSKPGV IFLTKRSRQV CADPSEEWVQ KYVSDLELSA | |
Greater than 95% as determined by SDS-PAGE. |
6348 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human CCL3 / Mip1a induces THP-1 chemotaxis. Activity was further confirmed by measuring Human PBMC chemotaxis. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |