missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL5 / RANTES Protein
Recombinant Protein
Marque: enQuireBio™ QP5261-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_002976.2 | |
CCL5 / RANTES Protein | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the THP-1 chemotaxis assay. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
6352 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
CCL5 | |
Recombinant Protein | |
E. coli | |
SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS |