missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD164 Protein
A cDNA sequence encoding the CD164 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11305-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
8763 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPDKNTTQH PNVTTLAPIS NVTSAPVTSL PLVTTPAPET CEGRNSCVSC FNVSVVNTTC FWIECKDESY CSHNSTVSDC QVGNTTDFCS VSTATPVPTA NSTAKPTVQP SPSTTSKTVT TSGTTNNTVT PTSQPVRKST FDHHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |
CD164 Protein | |
Research Use Only | |
CD164 | |
Human | |
His | |
CD164 protein solution (0.5 mg/ml) contains 10% glycerol and Phosphate buffered saline (pH 7.4). |