missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD1B Protein
A cDNA sequence encoding the CD1B was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11309-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
910 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEHAFQ GPTSFHVIQT SSFTNSTWAQ TQGSGWLDDL QIHGWDSDSG TAIFLKPWSK GNFSDKEVAE LEEIFRVYIF GFAREVQDFA GDFQMKYPFE IQGIAGCELH SGGAIVSFLR GALGGLDFLS VKNASCVPSP EGGSRAQKFC ALIIQYQGIM ETVRILLYET CPRYLLGVLN AGKADLQRQV KPEAWLSSGP SPGPGRLQLV CHVSGFYPKP VWVMWMRGEQ EQQGTQLGDI LPNANWTWYL RATLDVADGE AAGLSCRVKH SSLEGQDIIL YWRNPTSIGS | |
Greater than 90.0% as determined by SDS-PAGE. |
CD1B Protein | |
Research Use Only | |
CD1B | |
Human | |
His | |
CD1B protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 20% glycerol and 1mM DTT. |