Learn More
enQuireBio™ Recombinant Human CEACAM7 Protein
A DNA sequence encoding the Homo sapiens (Human) CEACAM7, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5814-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001278414.1 | |
CEACAM7 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1087 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CEACAM7 | |
Recombinant Protein | |
E. coli | |
TNIDVVPFNV AEGKEVLLVV HNESQNLYGY NWYKGERVHA NYRIIGYVKN ISQENAPGPA HNGRETIYPN GTLLIQNVTH NDAGFYTLHV IKENLVNEEV TRQFYVFSEP PKPSITSNNF NPVENKDIVV LTCQPETQNT TYLWWVNNQS LLVSPRLLLS TDNRTLVLLS ATKNDIGPYE CEIQNPVGAS RSDPVTLNVR YESVQAS |