missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cementoblastoma-derived protein 1
A DNA sequence encoding the Homo sapiens (Human) Cementoblastoma-derived protein 1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP9821-ye-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001041677.1 | |
Cementoblastoma-derived protein 1 | |
Research Use Only | |
CEMP1 | |
Recombinant Protein | |
Yeast | |
MGTSSTDSQQ AGHRRCSTSN TSAENLTCLS LPGSPGKTAP LPGPAQAGAG QPLPKGCAAV KAEVGIPAPH TSQEVRIHIR RLLSWAAPGA CGLRSTPCAL PQALPQARPC PGRWFFPGCS LPTGGAQTIL SLWTWRHFLN WALQQREENS GRARRVPPVP RTAPVSKGEG SHPPQNSNGE KVKTITPDVG LHQSLTSDPT VAVLRAKRAP EAHPPRSCSG SLTARVCHMG VCQGQGDTED GRMTLMG | |
Greater than 90% as determined by SDS-PAGE. |
752014 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |