Learn More
enQuireBio™ Recombinant Human CKB Protein
A cDNA sequence encoding the CKB was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP11435-10ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15997248
  • 203.00€ / 10µg
Expédition estimée: 10-05-2024
Spécification
1152
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MPFSNSHNAL KLRFPAEDEF PDLSAHNNHM AKVLTPELYA ELRAKSTPSG FTLDDVIQTG VDNPGHPYIM TVGCVAGDEE SYEVFKDLFD PIIEDRHGGY KPSDEHKTDL NPDNLQGGDD LDPNYVLSSR VRTGRSIRGF CLPPHCSRGE RRAIEKLAVE ALSSLDGDLA GRYYALKSMT EAEQQQLIDD HFLFDKPVSP LLLASGMARD WPDARGIWHN DNKTFLVWVN EEDHLRVISM QKGGNMKEVF TRFCTGLTQI ETLFKSKDYE FMWNPHLGYI LTCPSNLGTG LRAGVHIKLP NLGKHEKFSE VLKRLRLQKR GTGGVDTAAV GGVFDVSNAD RLGFSEVELV QMVVDGVKLL IEMEQRLEQG QAIDDLMPAQ K
Greater than 95.0% as determined by SDS-PAGE.
CKB Protein
Research Use Only
CKB
Human
Untagged
CKB Human solution containing 20mM Trsi pH 8.0, 1mM DTT, and 10% glycerol.