missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human COPE Protein

A cDNA sequence encoding the COPE was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11484-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15927408

  • 137.00€ / 2µg
Expédition estimée: 27-06-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

11316
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
COPE
Human
His
COPE protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT.
COPE Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAPPAPG PASGGSGEVD ELFDVKNAFY IGSYQQCINE AQRVKLSSPE RDVERDVFLY RAYLAQRKFG VVLDEIKPSS APELQAVRMF ADYLAHESRR DSIVAELDRE MSRSVDVTNT TFLLMAASIY LHDQNPDAAL RALHQGDSLE CTAMTVQILL KLDRLDLARK ELKRMQDLDE DATLTQLATA WVSLATGGEK LQDAYYIFQE MADKCSPTLL LLNGQAACHM AQGRWEAAEG LLQEALDKDS GYPETLVNLI VLSQHLGKPP EVTNRYLSQL KDAHRSHPFI KEYQAKENDF DRLVLQYAPS A
Vidéos
FDS
Documents
Certificats
Promotions

Promotions