Learn More
enQuireBio™ Recombinant Human CSF2RB Protein
A cDNA sequence encoding the CSF2RB was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11529-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
CSF2RB Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CSF2RB protein solution (0.5 mg/ml) containing Phosphate Buffered Saline(pH 7.4) and 10% glycerol. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
WERSLAGAEE TIPLQTLRCY NDYTSHITCR WADTQDAQRL VNVTLIRRVN EDLLEPVSCD LSDDMPWSAC PHPRCVPRRC VIPCQSFVVT DVDYFSFQPD RPLGTRLTVT LTQHVQPPEP RDLQISTDQD HFLLTWSVAL GSPQSHWLSP GDLEFEVVYK RLQDSWEDAA ILLSNTSQAT LGPEHLMPSS TYVARVRTRL APGSRLSGRP SKWSPEVCWD SQPGDEAQPQ NLECFFDGAA VLSCSWEVRK EVASSVSFGL FYKPSPDAGE EECSPVLREG LGSLHTRHHC QIPVPDPATH GQYIVSVQPR RAEKHIKSSV NIQMAPPSLN VTKDGDSYSL RWETMKMRYE HIDHTFEIQY RKDTATWKDS KTETLQNAHS MALPALEPST RYWARVRVRT SRTGYNGIWS EWSEARSWDT ESVLPMWLEH HHHHH |