Learn More
enQuireBio™ Recombinant Human CST11 Protein
A cDNA sequence encoding the CST11 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11536-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
CST11 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CST11 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M urea and 10% glycerol. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSARKKTFL SVHEVMAVEN YAKDSLQWIT DQYNKESDDK YHFRIFRVLK VQRQQVNCFF SVFAVPWFEQ YKILNKSCSS D |