missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTDSP1 Protein
A cDNA sequence encoding the CTDSP1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11550-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
CTDSP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDSSAVITQI SKEEARGPLR GKGDQKSAAS QKPRSRGILH SLFCCVCRDD GEALPAHSGA PLLVEENGAI PKTPVQYLLP EAKAQDSDKI CVVIDLDETL VHSSFKPVNN ADFIIPVEID GVVHQVYVLK RPHVDEFLQR MGELFECVLF TASLAKYADP VADLLDKWGA FRARLFRESC VFHRGNYVKD LSRLGRDLRR VLILDNSPAS YVFHPDNAVP VASWFDNMSD TELHDLLPFF EQLSRVDDVY SVLRQPRPGS | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CTDSP1 solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 20% glycerol and 0.1M NaCl. |