missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cyclophilin G Protein

A cDNA sequence encoding the Cyclophilin G was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10574-25ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15994288

  • 203.00€ / 25µg
Expédition estimée: 11-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

9360
25 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 200 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.
Human
His
Cyclophilin-G solution containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.
Cyclophilin G Protein
Research Use Only
PPIG
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGIKVQRPRC FFDIAINNQP AGRVVFELFS DVCPKTCENF RCLCTGEKGT GKSTQKPLHY KSCLFHRVVK DFMVQGGDFS EGNGRGGESI YGGFFEDESF AVKHNKEFLL SMANRGKDTN GSQFFITTKP TPHLDGHHVV FGQVISGQEV VREIENQKTD AASKPFAEVR ILSCG
Greater than 95.0% as determined by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions