Learn More
Novus Biologicals™ Recombinant Human Cyr61/CCN1 Protein
Code produit. 18478651 Tous les produits Bio Techne Produits
Click to view available options
Quantité:
1 mg
100 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
500µg
5µg
Code produit. 18478651
Marque: Novus Biologicals™ NBP2349441mg
Recombinant Protein
Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine Balb/3T3 cells is less than 3.0 μg/mL, corresponding to a specific activity of >330 IU/mg.
TRUSTED_SUSTAINABILITY
Spécification
Numéro d’adhésion O00622
Concentration LYOPH
À utiliser avec (application) PAGE, Bioactivity
Formule Lyophilized from a 0.2 um filtered concentrated solution in citrate buffer solution, 300 mM NaCl, pH 3.0.
Identification génétique (Entrez) 3491
Poids moléculaire 39.4 kDa
Quantité 1 mg
Conditions de stockage Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Concentration en endotoxines Less than 1 EU/ug of Cyr61/CCN1 as determined by LAL method.
Alias de gène CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
Symbole de gène(s) CCN1
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Catégorie de recherche Biologically Active Proteins
Pureté ou qualité >95% by SDS-PAGE and HPLC
Protéine Cyr61/CCN1