missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Cyr61/CCN1 Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
1 mg
100 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
500µg
5µg
Description
Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine Balb/3T3 cells is less than 3.0 μg/mL, corresponding to a specific activity of >330 IU/mg.
Spécification
Spécification
| Numéro d’adhésion | O00622 |
| Concentration | LYOPH |
| À utiliser avec (application) | PAGE, Bioactivity |
| Formule | Lyophilized from a 0.2 um filtered concentrated solution in citrate buffer solution, 300 mM NaCl, pH 3.0. |
| Identification génétique (Entrez) | 3491 |
| Poids moléculaire | 39.4 kDa |
| Quantité | 100 μg |
| Conditions de stockage | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Concentration en endotoxines | Less than 1 EU/ug of Cyr61/CCN1 as determined by LAL method. |
| Alias de gène | CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu