Learn More
enQuireBio™ Recombinant Human Cytokeratin 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 10, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6275-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_000412.3 | |
Cytokeratin 10 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3858 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
KRT10 | |
Recombinant Protein | |
E. coli | |
MSVRYSSSKH YSSSRSGGGG GGGGCGGGGG VSSLRISSSK GSLGGGFSSG GFSGGSFSRG SSGGGCFGGS SGGYGGLGGF GGGSFRGSYG SSSFGGSYGG IFGGGSFGGG SFGGGSFGGG GFGGGGFGGG FGGGFGGDGG LLSGNEKVTM QNLNDRLASY LDKVRALEES NYELEGKIKE WYEKHGNSHQ GEPRDYSKYY KTIDDLKNQI LNLTTDNANI LLQIDNARLA ADDFRLKYEN EVALRQSVEA DINGLRRVLD ELTLTKADLE MQIESLTEEL AYLKKNHEEE MKDLRNVSTG DVNVEMNAAP GVDLTQLLNN MRSQYEQLAE QNRKDAEAWF NEKSKELTTE IDNNIEQISS YKSEITELRR NVQALEIELQ SQLALKQSLE ASLAETEGRY CVQLSQIQAQ ISALEEQLQQ IRAETECQNT EYQQLLDIKI RLENEIQTYR SLLEGEGSSG GGGRGGGSFG GGYGGGSSGG GSSGGGHGGG HGGSSGGGYG GGSSGGGSSG GGYGGGSSSG GHGGSSSGGY GGGSSGGGGG GYGGGSSGGG SSSGGGYGGG SSSGGHKSSS SGSVGESSSK GPRY |