missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DDT Protein
A cDNA sequence encoding the DDT was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11612-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
1652 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPFLELDTNL PANRVPAGLE KRLCAAAASI LGKPADRVNV TVRPGLAMAL SGSTEPCAQL SISSIGVVGT AEDNRSHSAH FFEFLTKELA LGQDRILIRF FPLESWQIGK IGTVMTFL | |
Greater than 95.0% as determined by SDS-PAGE. |
DDT Protein | |
Research Use Only | |
DDT | |
Human | |
His | |
DDT Human solution containing 20mM Tris-HCl pH 8.0, and 10% glycerol. |