Learn More
enQuireBio™ Recombinant Human Dermcidin / DCD Protein
A DNA sequence encoding the Homo sapiens (Human) Dermcidin / DCD, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5914-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001287783.1 | |
Dermcidin / DCD Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
117159 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
DCD | |
Recombinant Protein | |
E. coli | |
YDPEAASAPG SGNPCHEASA AQKENAGEDP GLARQAPKPR KQRSSLLEKG LDGAKKAVGG LGKLGKDAVE DLESVGKGAV HDVKDVLDSV L |