missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA-3-methyladenine glycosylase Protein
A DNA sequence encoding the Homo sapiens (Human) DNA-3-methyladenine glycosylase, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP9282-ye-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001015052.1 | |
DNA-3-methyladenine glycosylase Protein | |
Research Use Only | |
MPG | |
Recombinant Protein | |
Yeast | |
QFCRRMGQKK QRPARAGQPH SSSDAAQAPA EQPHSSSDAA QAPCPRERCL GPPTTPGPYR SIYFSSPKGH LTRLGLEFFD QPAVPLARAF LGQVLVRRLP NGTELRGRIV ETEAYLGPED EAAHSRGGRQ TPRNRGMFMK PGTLYVYIIY GMYFCMNISS QGDGACVLLR ALEPLEGLET MRQLRSTLRK GTASRVLKDR ELCSGPSKLC QALAINKSFD QRDLAQDEAV WLERGPLEPS EPAVVAAARV GVGHAGEWAR KPLRFYVRGS PWVSVVDRVA EQDTQA | |
Greater than 90% as determined by SDS-PAGE. |
4350 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |