missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA replication complex GINS protein PSF2 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA replication complex GINS protein PSF2, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP8265-ec-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_057179.1 | |
DNA replication complex GINS protein PSF2 Protein | |
Research Use Only | |
GINS2 | |
Recombinant Protein | |
E. coli | |
MDAAEVEFLA EKELVTIIPN FSLDKIYLIG GDLGPFNPGL PVEVPLWLAI NLKQRQKCRL LPPEWMDVEK LEKMRDHERK EETFTPMPSP YYMELTKLLL NHASDNIPKA DEIRTLVKDM WDTRIAKLRV SADSFVRQQE AHAKLDNLTL MEINTSGTFL TQALNHMYKL RTNLQPLEST QSQDF | |
Greater than 90% as determined by SDS-PAGE. |
51659 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |