Learn More
enQuireBio™ Recombinant Human EDN3 Protein
A cDNA sequence encoding the EDN3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11747-HIS-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
1908 | |
Greater than 97.0% as determined by RP-HPLC. | |
Research Use Only | |
EDN3 | |
Human | |
His | |
The protein (1 mg/ml) was lyophilized with no additives. |
EDN3 Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
H-Cys-Thr-Cys-Phe-Thr-Tyr-Lys-Asp-Lys-Glu-Cys-Val-Tyr-Tyr-Cys-His-Leu-Asp-Ile-Ile-Trp-OH |