missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EFNB1 Protein

A cDNA sequence encoding the EFNB1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11753-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15998248

  • 205.00€ / 20µg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

EFNB1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The EFNB1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 5% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MLAKNLEPVS WSSLNPKFLS GKGLVIYPKI GDKLDIICPR AEAGRPYEYY KLYLVRPEQA AACSTVLDPN VLVTCNRPEQ EIRFTIKFQE FSPNYMGLEF KKHHDYYITS TSNGSLEGLE NREGGVCRTR TMKIIMKVGQ DPNAVTPEQL TTSRPSKEAD NTVKMATQAP GSRGSLGDSD GKHETVNQEE KSGPGASGGS SGDPDGFFNS K
Vidéos
FDS
Documents
Certificats
Promotions

Promotions