missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EFNB2 Protein

A cDNA sequence encoding the EFNB2 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11754-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15918258

  • 4060.00€ / 1mg
Expédition estimée: 27-06-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

EFNB2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
EFNB2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 20% glycerol and 200mM NaCl.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMIVLEP IYWNSSNSKF LPGQGLVLYP QIGDKLDIIC PKVDSKTVGQ YEYYKVYMVD KDQADRCTIK KENTPLLNCA KPDQDIKFTI KFQEFSPNLW GLEFQKNKDY YIISTSNGSL EGLDNQEGGV CQTRAMKILM KVGQDASSAG STRNKDPTRR PELEAGTNGR SSTTSPFVKP NPGSSTDGNS AGHSGNNILG SEVALFA
Vidéos
FDS
Documents
Certificats
Promotions

Promotions