Learn More
enQuireBio™ Recombinant Human EGLN3 Protein
A cDNA sequence encoding the EGLN3 was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP11761-10ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15998278
  • 203.00€ / 10µg
Expédition estimée: 10-05-2024
Spécification
112399
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMPLGHI MRLDLEKIAL EYIVPCLHEV GFCYLDNFLG EVVGDCVLER VKQLHCTGAL RDGQLAGPRA GVSKRHLRGD QITWIGGNEE GCEAISFLLS LIDRLVLYCG SRLGKYYVKE RSKAMVACYP GNGTGYVRHV DNPNGDGRCI TCIYYLNKNW DAKLHGGILR IFPEGKSFIA DVEPIFDRLL FFWSDRRNPH EVQPSYATRY AMTVWYFDAE ERAEAKKKFR NLTRKTESAL TED
Greater than 90% as determined by SDS-PAGE.
EGLN3 Protein
Research Use Only
EGLN3
Human
His
The EGLN3 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 300mM NaCl, 5mM DTT, 2mM EDTA and 50% glycerol.