Learn More
enQuireBio™ Recombinant Human Ferritin Protein
A cDNA sequence encoding the Ferritin was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11867-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
Ferritin Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
The protein solution is in 0.05M TRIS buffer pH 7.5 containing 1.0M NaCl and 0.09% NaN3. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 96.0%. |