Learn More
enQuireBio™ Recombinant Human FGF10 / KGF2 Protein
Recombinant Protein
Marque: enQuireBio™ QP5311-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_004456.1 | |
FGF10 / KGF2 Protein | |
Research Use Only | |
FGF10 | |
Recombinant Protein | |
E. coli | |
MLGQDMVSPE . ATNSSSSSF S. SPSSAGRH VR. SYNHLQG DVR. WRKLFS FTKY. FLKIE KNGKV. SGTK KENCPY. SIL EITSVEIGVV AVKAINS. NY YLAMNKKG. K LYGSKEFNN. DCKLKERIEE . NGYNTYASF N. WQHNGRQM YV. ALNGKGA PRR. GQKTRR KNTS. AHFLP MVVHS | |
Greater than 95% as determined by SDS-PAGE. |
2255 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
4MBr-5 cell proliferation is significantly induced by less than 200 ng/ml of human FGF10 or activity measured must be greater than 5.0 x 10^3 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |