missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLTP Protein
A cDNA sequence encoding the GLTP was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11993-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
GLTP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMALLAEH LLKPLPADKQ IETGPFLEAV SHLPPFFDCL GSPVFTPIKA DISGNITKIK AVYDTNPAKF RTLQNILEVE KEMYGAEWPK VGATLALMWL KRGLRFIQVF LQSICDGERD ENHPNLIRVN ATKAYEMALK KYHGWIVQKI FQAALYAAPY KSDFLKALSK GQNVTEEECL EKIRLFLVNY TATIDVIYEM YTQMNAELNY KV | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GLTP protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 20% glycerol and 1mM DTT. |