Learn More
enQuireBio™ Recombinant Human GNG4 Protein
A cDNA sequence encoding the GNG4 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12019-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
GNG4 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GNG4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKEGMSN NSTTSISQAR KAVEQLKMEA CMDRVKVSQA AADLLAYCEA HVREDPLIIP VPASENPFRE KKFFC |