missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human GPAM GST (N-Term) Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
10 μg
25 μg
Conditionnement:
10µg
25µg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Especificaciones
Especificaciones
| À utiliser avec (application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formule | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Identification génétique (Entrez) | 57678 |
| Poids moléculaire | MolecularWeight-theroretical: 34.54 kDa |
| Quantité | 25 μg |
| Conditions de stockage | Store at -80°C. Avoid freeze-thaw cycles. |
| Alias de gène | EC 2.3.1, EC 2.3.1.15, glycerol-3-phosphate acyltransferase 1, mitochondrial, glycerol-3-phosphate acyltransferase, mitochondrial, GPAT, GPAT-1, GPAT1mitochondrial, MGC26846 |
| Symbole de gène(s) | GPAM |
| Catégorie de recherche | metabolism |
| Pureté ou qualité | >80% by SDS-PAGE and Coomassie blue staining |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido