missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HOMER3 Protein
A cDNA sequence encoding the HOMER3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12293-1mg
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01000kg
Spécification
HOMER3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMMSTAR EQPIFSTRAH VFQIDPATKR NWIPAGKHAL TVSYFYDATR NVYRIISIGG AKAIINSTVT PNMTFTKTSQ KFGQWADSRA NTVYGLGFAS EQHLTQFAEK FQEVKEAARL AREKSQDGGE LTSPALGLAS HQVPPSPLVS ANGPGEEKLF RSQSADAPGP TERERLKKML SEGSVGEVQW EAEFFALQDS NNKLAGALRE ANAAAAQWRQ QLEAQRAEAE RLRQRVAELE AQAASEVTPT GEKEGLGQGQ SLEQLEALVQ TKDQEIQTLK SQTGGPREAL EAAEREETQQ KVQDLETRNA ELEHQLRAME RSLEEARAER ERARAEVGRA AQLLDVSLFE LSELREGLAR LAEAAP | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HOMER3 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 150mM NaCl, 1mM DTT and 30% glycerol. |