Learn More
enQuireBio™ Recombinant Human HRASLS3 Protein
A cDNA sequence encoding the HRASLS3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12312-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
HRASLS3 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein contains 20mM Tris-HCl buffer pH 8. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRAPIPEPKP GDLIEIFRPF YRHWAIYVGD GYVVHLAPPS EVAGAGAASV MSALTDKAIV KKELLYDVAG SDKYQVNNKH DDKYSPLPCS KIIQRAEELV GQEVLYKLTS ENCEHFVNEL RYGVARSDQV RDV |