Learn More
enQuireBio™ Recombinant Human HSPB3 Protein
A cDNA sequence encoding the HSPB3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12332-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
HSPB3 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HSPB3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 5% glycerol. |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAKIILRHLI EIPVRYQEEF EARGLEDCRL DHALYALPGP TIVDLRKTRA AQSPPVDSAA ETPPREGKSH FQILLDVVQF LPEDIIIQTF EGWLLIKAQH GTRMDEHGFI SRSFTRQYKL PDGVEIKDLS AVLCHDGILV VEVKDPVGTK |