missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSPB7 Protein

A cDNA sequence encoding the HSPB7 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12334-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15980089

  • 189.00€ / 10µg
Expédition estimée: 10-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

HSPB7 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HSPB7 Human solution containing 20mM Tris HCl pH 8.0, 0.2M NaCl, 2mM DTT and 50% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSHRTSSTFR AERSFHSSSS SSSSSTSSSA SRALPAQDPP MEKALSMFSD DFGSFMRPHS EPLAFPARPG GAGNIKTLGD AYEFAVDVRD FSPEDIIVTT SNNHIEVRAE KLAADGTVMN TFAHKCQLPE DVDPTSVTSA LREDGSLTIR ARRHPHTEHV QQTFRTEIKI
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions