Learn More
enQuireBio™ Recombinant Human IER3 Protein
A cDNA sequence encoding the IER3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12370-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
8870 | |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
IER3 | |
Human | |
His | |
IER3 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |
IER3 Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMCHSRSC HPTMTILQAP TPAPSTIPGP RRGSGPEIFT FDPLPEPAAA PAGRPSASRG HRKRSRRVLY PRVVRRQLPV EEPNP |