missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGFBP 1 Protein
A cDNA sequence encoding the IGFBP 1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10683-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
IGFBP 1 Protein | |
Research Use Only | |
The ED50, as determined by the inhibition of rHuIGF-I-induced proliferation of human MCF-7 cells, is less than 4microg/ml. | |
Human | |
Untagged | |
IGFBP-1 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS. |
25 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human | |
APWQCAPCSA EKLALCPPVS ASCSEVTRSA GCGCCPMCAL PLGAACGVAT ARCARGLSCR ALPGEQQPLH ALTRGQGACV QESDASAPHA AEAGSPESPE STEITEEELL DNFHLMAPSE EDHSILWDAI STYDGSKALH VTNIKKWKEP CRIELYRVVE SLAKAQETSG EEISKFYLPN CNKNGFYHSR QCETSMDGEA GLCWCVYPWN GKRIPGSPEI RGDPNCQIYF NVQN | |
Greater than 95.0% as determined by SDS-PAGE. |