missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL 28A Protein

A cDNA sequence encoding the IL 28A was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10707-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15924928

  • 131.00€ / 2µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

282616
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
The specific activity was determined by the dose dependent protection of the cytopathic effect on A549 cells (human lung adenocarcinoma epithelial cell line) that were challenged with encephalomyocarditis (EMC) virus and is typically 0.5-5ng/ml.
Human
Untagged
The IL28A was lyophilized from 1 mg/ml in 1xPBS.
IL 28A Protein
Research Use Only
IFNL2
Recombinant Protein
Human (HEK293)
MKLDMTGDCT PVLVLMAAVL TVTGAVPVAR LHGALPDARG CHIAQFKSLS PQELQAFKRA KDALEESLLL KDCRCHSRLF PRTWDLRQLQ VRERPMALEA ELALTLKVLE ATADTDPALV DVLDQPLHTL HHILSQFRAC IQPQPTAGPR TRGRLHHWLY RLQEAPKKES PGCLEASVTF NLFRLLTRDL NCVASGDLCV
Greater than 95% as obsereved by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions