missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Integrin beta 1/CD29 Protein
Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantité:
250 μg
50 μg
Förpackningsstorlek
250µg
50µg
Beskrivelse
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-728 of Human Integrin beta 1/CD29 The Recombinant Human Integrin beta 1/CD29 Protein is derived from E. coli. The Recombinant Human Integrin beta 1/CD29 Protein has been validated for the following applications: SDS-Page.
Tekniske data
Tekniske data
| Concentration | 1mg/mL |
| À utiliser avec (application) | SDS-PAGE |
| Formule | Liquid. PBS (pH 7.4) containing 10% glycerol |
| Identification génétique (Entrez) | 3688 |
| Poids moléculaire | 79.4 kDa |
| Nom | Recombinant Human Integrin beta 1/CD29 Protein |
| Méthode de purification | >90%, by SDS-PAGE |
| Quantité | 50 μg |
| Source | Baculovirus |
| Conditions de stockage | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Vis mere |
For Research Use Only.
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion