missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human IRF9 GST (N-Term) Protein

Product Code. p-200053225 Shop All Bio Techne Products
Click to view available options
Quantité:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 18317479

Brand: Novus Biologicals™ H00010379P0110ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

À utiliser avec (application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formule 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Identification génétique (Entrez) 10379
Poids moléculaire 68.97 kDa
Quantité 10 μg
Conditions de stockage Store at -80°C. Avoid freeze-thaw cycles.
Alias de gène IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma
Symbole de gène(s) IRF9
Catégorie de recherche Signal Transduction
Pureté ou qualité >80% by SDS-PAGE and Coomassie blue staining
Protéine IRF9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.