missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IVD Protein

A cDNA sequence encoding the IVD was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12466-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15980509

  • 3120.00€ / 1mg
Expédition estimée: 12-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

3712
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
IVD
Human
His
The IVD protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.
IVD Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MHSLLPVDDA INGLSEEQRQ LRQTMAKFLQ EHLAPKAQEI DRSNEFKNLR EFWKQLGNLG VLGITAPVQY GGSGLGYLEH VLVMEEISRA SGAVGLSYGA HSNLCINQLV RNGNEAQKEK YLPKLISGEY IGALAMSEPN AGSDVVSMKL KAEKKGNHYI LNGNKFWITN GPDADVLIVY AKTDLAAVPA SRGITAFIVE KGMPGFSTSK KLDKLGMRGS NTCELIFEDC KIPAANILGH ENKGVYVLMS GLDLERLVLA GGPLGLMQAV LDHTIPYLHV REAFGQKIGH FQLMQGKMAD MYTRLMACRQ YVYNVAKACD EGHCTAKDCA GVILYSAECA TQVALDGIQC FGGNGYINDF PMGRFLRDAK LYEIGAGTSE VRRLVIGRAF NADFH
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions