missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIF Protein
Recombinant Protein
Marque: enQuireBio™ QP2835-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001244064.1 | |
LIF Protein | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 200 pg/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 5 x 10^6 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
3976 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
LIF | |
Recombinant Protein | |
E. coli | |
MSPLPITPVN ATCAIRHPCH NNLMNQIRSQ LAQLNGSANA LFILYYTAQG EPFPNNLDKL CGPNVTDFPP FHANGTEKAK LVELYRIVVY LGTSLGNITR DQKILNPSAL SLHSKLNATA DILRGLLSNV LCRLCSKYHV GHVDVTYGPD TSGKDVFQKK KLGCQLLGKY KQIIAVLAQA F |